Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Goat anti-Canine IgG (H+L) Secondary Antibody, FITC, Invitrogen™
Goat Polyclonal Secondary Antibody
Supplier: Thermo Scientific SA510281

Description
This antibody will react with dog IgG and with light chains common with other dog immunoglobulins as determined by ELISA and IEP techniques. This antibody may cross-react with IgG from other species.
Canine IgG (H+L) Polyclonal antibody specifically detects Canine IgG (H+L) in Canine samples. It is validated for ELISA, Immunocytochemistry, Immunoelectrophoresis, Western Blot.
Specifications
Canine IgG (H+L) | |
Polyclonal | |
FITC | |
Goat | |
Antigen affinity chromatography | |
RUO | |
Canine | |
Liquid |
ELISA, Immunocytochemistry, Immunoelectrophoresis, Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Canine IgG h+l | |
1 mg | |
Secondary | |
4° C, store in dark | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction