Learn More
Goat anti-Chicken IgA Heavy Chain Secondary Antibody, FITC, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | IgA Heavy Chain |
| Applications | Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | FITC |
| Dilution | Immunohistochemistry 1:50-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:500, Immunohistochemistry-Paraffin 1:50 - 1:500 |
| Gene Alias | Immunoglobulin A |
| Gene Symbols | IGHA1 |
| Host Species | Goat |
| Immunogen | Chicken IgA |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.