Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Chicken IgM (mu), HRP, Polyclonal, Secondary Antibody, Abnova™

Catalog No. 89138532 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-532 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-532 Supplier Abnova Corporation Supplier No. PAB29165

Specifications

Antigen Goat anti-Chicken IgM (mu)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate HRP
Description Goat anti-chicken IgM recognizes chicken IgM (mu) . The anti-mu-specific fraction was purified using antigen affinity chromatography.
Dilution ELISA (1:10000) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation Liquid
Host Species Goat
Immunogen Native purified chicken IgM from normal chicken serum.
Purification Method Affinity purification
Quantity 500 μg
Primary or Secondary Secondary
Target Species Chicken
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.