Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat Anti-Chicken IgY Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. AF010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF010 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF010 Supplier Novus Biologicals Supplier No. AF010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IgY Polyclonal specifically detects IgY in Chicken samples. It is validated for Western Blot.

Specifications

Antigen IgY
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:5000 dilution
Host Species Goat
Immunogen Chicken IgY
Purification Method Affinity Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity Detects chicken IgY heavy and light chains. In direct ELISA, less than 1% cross-reactivity with human IgG, rabbit IgG, rat IgG, donkey IgG and mouse IgG is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Chicken
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.