Learn More
Goat anti-Chicken IgY Fab, Polyclonal, Secondary Antibody, Invitrogen™
Goat Polyclonal Secondary Antibody
Supplier: Thermo Scientific SA172005
Description
Thermo Scientific Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
SA1-72005 detects Chicken IgY Fab in chicken samples.SA1-72005 has been successfully used in ELISA and western blot procedures.The SA1-72005 immunogen is from purified chicken IgY Fab fragments isolated from normal chicken serum.
Specifications
Chicken IgY Fab | |
Polyclonal | |
Unconjugated | |
Purified chicken IgY Fab fragments isolated from normal chicken serum. | |
500 μg | |
Secondary | |
Store at 4°C | |
IgG |
ELISA, Western Blot | |
1mg/mL | |
Goat | |
Affinity chromatography | |
RUO | |
Chicken | |
Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.