Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Chicken IgY Fc Secondary Antibody, DyLight 350, R&D Systems™
SDP

Catalog No. NB538143 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB538143 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB538143 Supplier R&D Systems Supplier No. NB7286UV
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Goat Polyclonal Antibody

IgY Fc Polyclonal specifically detects IgY Fc in Chicken samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen IgY Fc
Applications ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Host Species Goat
Immunogen Chicken IgG (IgY)-Fc Fragment
Purification Method Immunogen affinity purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Chicken
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.