Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Chicken IgY, HRP, Polyclonal, Secondary Antibody, Abnova™

Catalog No. 89138475 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-475 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-475 Supplier Abnova Corporation Supplier No. PAB29099
Add to Cart
Add to Cart

Specifications

Antigen Goat anti-Chicken IgY
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate HRP
Description Goat polyclonal antibody raised against Chicken IgY. The antibody is conjugated with HRP.
Dilution ELISA Immunocytochemistry Immunohistochemistry Western Blot(1:5000) The optimal working dilution should be determined by the end user.
Formulation Liquid
Host Species Goat
Immunogen Purified chicken IgY (whole protein) emulsified in Freund's adjuvant.
Purification Method Affinity purification
Quantity 400 μL
Primary or Secondary Secondary
Target Species Chicken
Content And Storage Store at 4°C and avoid from light. For long term storage dilute the antibody with sterile glycerol and store it at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.