Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Feline IgG Fc Secondary Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NBP173499 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP173499 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP173499 Supplier Novus Biologicals Supplier No. NBP173499
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG Fc Polyclonal specifically detects IgG Fc in Feline samples. It is validated for ELISA, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG Fc
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000 - 1:5000
Host Species Goat
Immunogen Cat IgG F(c) fragment
Purification Method Affinity Purified
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity No reaction was observed against Cat IgG F(ab).
Reconstitution Reconstitute with 1.0 ml deionized water (or equivalent).
Target Species Feline
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.