Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Goat anti-Feline IgG (H+L) Secondary Antibody, Novus Biologicals™

Goat Polyclonal Antibody
$220.50
Specifications
Antigen | IgG (H+L) |
---|---|
Concentration | 4.5mg/mL |
Dilution | Flow Cytometry 1:20-1:2000, Immunomicroscopy 1:20-1:2000 |
Applications | Flow Cytometry, Immunomicroscopy |
Classification | Polyclonal |
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Feline samples. It is validated for Flow Cytometry, Immunomicroscopy.Specifications
IgG (H+L) | |
Flow Cytometry 1:20-1:2000, Immunomicroscopy 1:20-1:2000 | |
Polyclonal | |
Goat | |
10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.05% Sodium Azide | |
Secondary | |
Store at 4°C. |
4.5mg/mL | |
Flow Cytometry, Immunomicroscopy | |
Unconjugated | |
Adaptive Immunity, Immunology | |
IgG | |
Affinity purified | |
Based on IEP, this antibody reacts with heavy gamma chains on cat IgG and light chains on all cat immunoglobulins |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title