Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Goat anti-Ferret IgM Heavy Chain Secondary Antibody, FITC, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP173390
Description
IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Ferret samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA.Specifications
IgM Heavy Chain | |
Polyclonal | |
Fluorescein | |
Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain | |
Goat | |
Affinity Purified | |
RUO | |
3507 | |
Reconstitute with 1.0 ml deionized water (or equivalent). | |
Store at 4C in the dark. |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
LYOPH | |
Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
IGHM | |
Ferret IgM u mu chain | |
1 mg | |
Secondary | |
No reaction was observed against other ferret heavy or light chain proteins. | |
Ferret | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction