Learn More
Goat anti-Golden Syrian Hamster IgG (H+L) Secondary Antibody, HRP, R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | HRP |
| Dilution | Western Blot 1:1000 - 1:5000, ELISA 1:7000 - 1:35000, Immunohistochemistry 1:500 - 1:2500 |
| Host Species | Goat |
| Immunogen | Goat anti-Golden Syrian Hamster IgG (H+L) Secondary Antibody [HRP] was produced by repeated immunization with Golden Syrian Hamster IgG whole molecule in goat. |
| Purification Method | Affinity Purified |
| Quantity | 2 mg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.