Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Guinea Pig IgG F(ab')2 Secondary Antibody, HRP, R&D Systems™
SDP

Catalog No. NBP173519 Shop All Novus Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP173519 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP173519 Supplier R&D Systems Supplier No. NBP173519
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Goat Polyclonal Antibody

IgG F(ab')2 Polyclonal specifically detects IgG F(ab')2 in Guinea Pig samples. It is validated for ELISA, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG F(ab')2
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate HRP
Dilution Western Blot 1:1000 - 1:10000, ELISA 1:2000 - 1:6000, Immunohistochemistry 1:500 - 1:2500
Gene Alias immunoglobulin heavy constant gamma 1 (G1m marker)
Host Species Goat
Immunogen Guinea Pig IgG F(ab')2 fragment
Purification Method Affinity Purified
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity No reaction was observed against Guinea Pig IgG F(c).
Reconstitution Reconstitute with 1.0 ml deionized water (or equivalent).
Target Species Guinea Pig
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Isotype IgG
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.