Learn More
Goat anti-Guinea Pig IgG (H+L) Secondary Antibody, FITC (Pre-adsorbed), Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Dot Blot, Flow Cytometry, Fluorophore-Linked Immunosorbent Assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | FITC |
| Dilution | Flow Cytometry 1:500 - 1:2500, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Immunohistochemistry-Paraffin 1:200 - 1:500, Dot Blot, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 |
| Formulation | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with No Preservative |
| Host Species | Goat |
| Immunogen | Guinea Pig IgG whole molecule |
| Purification Method | Multi-step |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.