Learn More
Description
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | ELISA, Flow Cytometry, Immunoassay, Immunocytochemistry, Immunocytochemistry/Immunofluorescence, Immunodiffusion, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | PE |
| Formulation | PBS |
| Host Species | Goat |
| Immunogen | This Goat anti-Guinea Pig IgG (H+L) Secondary Antibody (Pre-adsorbed) was developed against purified guinea pig IgG (H&L). |
| Purification Method | Affinity purified |
| Quantity | 0.5 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
