Learn More
Goat anti-Guinea pig IgG, Whole Molecule, HRP, Polyclonal Secondary Antibody, MP Biomedicals
Shop All MP Biomedicals, Inc ProductsDescription
Host Species: Goat
Species Reactivity: Gp
Enzyme Immunoassay, Immunohistochemistry, Western Blotting, Electron Microscopy
Specifications
Specifications
| Antigen | IgG |
| Classification | Polyclonal |
| Concentration | 0.4mg/mL |
| Conjugate | HRP |
| Host Species | Goat |
| Purification Method | Affinity Purified |
| Quantity | 2 mL |
| Regulatory Status | RUO |
| Primary or Secondary | Secondary |
| Target Species | Guinea Pig |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.