Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Goat Anti-Human Biotinylated IgG, 4ml

Catalog No. PRV7830 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV7830 4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV7830 Supplier Promega Supplier No. V7830
May include imposed supplier surcharges.
Add to Cart
Add to Cart

High Capacity Magne Streptavidin Beads are magnetic affinity beads with high specificity and high capacity for binding biotinylated antibodies and proteins. Goat Anti-Human Biotinylated IgG is 0.5mg/ml in PBS (pH 7.4) with 0.1% sodium azide.

  • Enrich Antibody from Sera or Plasma with Magnetic Streptavidin Beads
  • Characterize a variety of antibody concentrations with a small amount of beads
  • Suitable for use with robotics and high−throughput platforms
  • Goat Anti−Human Biotinylated IgG is available separately

Specifications

Content And Storage 2°C to 10°C
Quantity 4 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.