Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Human IgA/IgG/IgM (H+L) Secondary Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NBP175041 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP175041 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP175041 Supplier Novus Biologicals Supplier No. NBP175041
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgA/IgG/IgM (H+L) Polyclonal specifically detects IgA/IgG/IgM (H+L) in Human samples. It is validated for Flow Cytometry, Immunomicroscopy.

Specifications

Antigen IgA/IgG/IgM (H+L)
Applications Flow Cytometry, Immunomicroscopy
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate FITC
Dilution Flow Cytometry 1:20-1:2000, Immunomicroscopy 1:20-1:2000
Host Species Goat
Immunogen Purified Human IgA + IgG + IgM
Purification Method Affinity purified
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity Based on IEP, this antibody reacts with heavy chains on human IgA + IgG + IgM and light chains on all human immunoglobulins
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgA
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.