Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Human IgD Heavy Chain Secondary Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB551261 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB551261 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB551261 Supplier Novus Biologicals Supplier No. NBP268528R
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgD Heavy Chain Polyclonal specifically detects IgD Heavy Chain in Human samples. It is validated for ELISA.

Specifications

Antigen IgD Heavy Chain
Applications ELISA
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias Constant Region of Heavy Chain of IgD, Ig Delta Chain C Region, Ig Delta Chain C Region NIG-65, Ig Delta Chain C Region WAH, Immunoglobulin Delta, immunoglobulin heavy constant delta
Host Species Goat
Immunogen Human IgD paraproteins
Purification Method Affinity purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.