Learn More
Immunology Consultants HRP Conjugated Goat anti-Human IgE Antibody, 1.0 mg
SDP
Supplier:  Immunology Consultants GE80P
Encompass_Preferred
Goats were immunized with highly purified human IgE and the resulting antiserum was collected. Antibody was solid phase adsorbed, immunoaffinity purified off a human IgE containing immunosorbent and conjugated with horseradish peroxidase (HRP). Antibody concentration was determined prior to conjugation using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with human IgE as determined by IEP and ELISA techniques. It is suitable for blotting, ELISA, and IHC applications. This antibody may cross-react with IgE from other species. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-196-2184
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.