Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Goat anti-Human IgE Heavy Chain Secondary Antibody, DyLight 550, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP175775
Description
IgE Heavy Chain Polyclonal antibody specifically detects IgE Heavy Chain in Human samples. It is validated for Flow Cytometry, Immunomicroscopy.Specifications
IgE Heavy Chain | |
Polyclonal | |
DyLight 550 | |
Constant Region of Heavy Chain of IgE, Ig Epsilon Chain C Region, Ig Epsilon Chain C Region ND, IgE, Immunoglobulin Epsilon, immunoglobulin heavy constant epsilon | |
Goat | |
Affinity purified | |
RUO | |
3497 | |
Rehydrate with 1.1 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. | |
Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage. |
Flow Cytometry, Immunomicroscopy | |
LYOPH | |
Flow Cytometry 1:20 - 1:2000, Immunomicroscopy 1:20 - 1:2000 | |
IGHE | |
Purified human IgE, (e chain) | |
1 mg | |
Secondary | |
Based on IEP, this antibody reacts with heavy epsilon chains on human IgE | |
Human | |
IgE |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction