Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Human IgE Heavy Chain Secondary Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP175778 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP175778 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP175778 Supplier Novus Biologicals Supplier No. NBP175778
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgE Heavy Chain Polyclonal specifically detects IgE Heavy Chain in Human samples. It is validated for Flow Cytometry, Immunomicroscopy.

Specifications

Antigen IgE Heavy Chain
Applications Flow Cytometry, Immunomicroscopy
Classification Polyclonal
Concentration LYOPH
Conjugate DyLight 650
Dilution Flow Cytometry 1:20 - 1:2000, Immunomicroscopy 1:20 - 1:2000
Gene Alias Constant Region of Heavy Chain of IgE, Ig Epsilon Chain C Region, Ig Epsilon Chain C Region ND, IgE, Immunoglobulin Epsilon, immunoglobulin heavy constant epsilon
Gene Symbols IGHE
Host Species Goat
Immunogen Purified human IgE, (e chain)
Purification Method Affinity purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3497
Test Specificity Based on IEP, this antibody reacts with heavy epsilon chains on human IgE
Reconstitution Rehydrate with 1.1 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily.
Target Species Human
Content And Storage Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage.
Isotype IgE
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.