Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Human IgE Heavy Chain Secondary Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB483716 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB483716 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB483716 Supplier Novus Biologicals Supplier No. NBP174923PCP
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgE Heavy Chain Polyclonal specifically detects IgE Heavy Chain in Human samples. It is validated for ELISA, Immunoassay, Immunodiffusion, Immunohistochemistry, Immunoprecipitation, Western Blot.

Specifications

Antigen IgE Heavy Chain
Applications ELISA, Immunoassay, Immunodiffusion, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias Constant Region of Heavy Chain of IgE, Ig Epsilon Chain C Region, Ig Epsilon Chain C Region ND, IgE, Immunoglobulin Epsilon, immunoglobulin heavy constant epsilon
Host Species Goat
Immunogen Purified human IgE, (Epsilon chain)
Purification Method Affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.