Learn More
Goat anti-Human IgE Heavy Chain Secondary Antibody (Pre-adsorbed), R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgE Heavy Chain |
| Applications | ELISA, Immunoassay, Immunodiffusion, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 4.5mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunoassay |
| Gene Alias | Constant Region of Heavy Chain of IgE, Ig Epsilon Chain C Region, Ig Epsilon Chain C Region ND, IgE, Immunoglobulin Epsilon, immunoglobulin heavy constant epsilon |
| Gene Symbols | IGHE |
| Host Species | Goat |
| Immunogen | Purified Human IgE |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.