Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Human IgG Fc Secondary Antibody (Pre-adsorbed), Novus Biologicals™
SDP

Catalog No. NBP260678 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP260678 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP260678 Supplier Novus Biologicals Supplier No. NBP260678
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG Fc Polyclonal specifically detects IgG Fc in Human samples. It is validated for Dot Blot, ELISA, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG Fc
Applications Dot Blot, ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, ELISA 1:15000-1:1:60000, Immunohistochemistry 1:1000-1:3000, Dot Blot
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Host Species Goat
Immunogen Human IgG F(c) fragment
Purification Method Affinity Purified
Quantity 1 mg
Primary or Secondary Secondary
Test Specificity This antibody was pre-adsorbed against Mouse Serum Proteins. No reaction was observed against Human IgG F(ab) or Mouse Serum Proteins. Specificity was confirmed by ELISA minimal cross reactivity against Mouse IgG.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.