Learn More
Goat anti-Human IgG Fraction, Whole Molecule Polyclonal Secondary Antibody, HRP, MP Biomedicals
Shop All MP Biomedicals, Inc ProductsDescription
Host Species: Goat
Isotype: IgG
Species Reactivity: Human
Specifications
Specifications
| Antigen | IgG, Whole Molecule |
| Classification | Polyclonal |
| Conjugate | HRP |
| Host Species | Goat |
| Quantity | 2 mL |
| Primary or Secondary | Secondary |
| Target Species | Human |
| Form | Purified |
| Isotype | IgG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.