Learn More
Goat anti-Human IgG (H+L) Secondary Antibody [Allophycocyanin], R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Dot Blot |
| Classification | Polyclonal |
| Conjugate | APC |
| Formulation | PBS |
| Host Species | Goat |
| Immunogen | Goat anti-Human IgG (H+L) Secondary Antibody was produced by repeated immunization with Human IgG whole molecule in goat. |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.5 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.