Learn More
Goat anti-Human IgG (H+L) Secondary Antibody, PE (Pre-adsorbed), R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Flow Cytometry, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | PE |
| Dilution | Flow Cytometry, Immunocytochemistry/Immunofluorescence 1:100 - 1:250 |
| Host Species | Goat |
| Immunogen | Goat anti-Human IgG (H+L) Secondary Antibody [PE] (Pre-adsorbed) was produced by repeated immunization with Human IgG whole molecule in goat. |
| Purification Method | Affinity Purified |
| Quantity | 0.5 mL |
| Regulatory Status | RUO |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.