Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Human IgG (H+L) Secondary Antibody [PerCP], Novus Biologicals™
SDP

Catalog No. NB785032 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB785032 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB785032 Supplier Novus Biologicals Supplier No. NBP260658PCP
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Dot Blot

Specifications

Antigen IgG (H+L)
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Dot Blot
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Host Species Goat
Immunogen Goat anti-Human IgG (H+L) Secondary Antibody was produced by repeated immunization with Human IgG whole molecule in goat.
Purification Method Immunogen affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.