Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Human IgG/IgM (H+L) Secondary Antibody, Novus Biologicals™
SDP

Catalog No. NBP266681 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266681 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266681 Supplier Novus Biologicals Supplier No. NBP266681
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG/IgM(H+L) Polyclonal specifically detects IgG/IgM(H+L) in Human samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG/IgM(H+L)
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:30000, ELISA 1:1000 - 1:30000, Immunohistochemistry 1:200 - 1:2000, Immunocytochemistry 1:200 - 1:2000
Formulation PBS with 0.09% Sodium Azide
Host Species Goat
Immunogen Human IgG+IgM-heavy and light chain
Purification Method Affinity Purified
Quantity 1 mL
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.