Learn More
Columbia Biosciences Corp GOAT ANTI-HUMAN IGGFC HRP 1MG
Supplier:  Columbia Biosciences Corp HRP110FC
Encompass_Preferred
Goat anti-Human IgG Fc-specific, secondary antibody, conjugated to HRP, 1mg
Catalog No. 50-1077-606
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.