Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Human IgM Heavy Chain Secondary Antibody [Janelia Fluor 525], Novus Biologicals™
SDP

Catalog No. NB785425 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB785425 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB785425 Supplier Novus Biologicals Supplier No. NBP175017JF525
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay

Specifications

Antigen IgM Heavy Chain
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay (CIA)
Classification Polyclonal
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain
Host Species Goat
Immunogen Purified Human IgM, Mu chain
Purification Method Affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3507
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.