Learn More
Goat anti-Human IgM Secondary Antibody, Invitrogen™
Description
Product # 31124 has been successfully used in Western blot, IF, ICC, IHC, ELISA, and ISH applications. The activity measured by a solid phase binding assay is 91%. Product # 31124 reacts with the μ chains of human IgM. Based on solid-phase enzyme immunoassay, this antibody cross-reacts with dog IgM (12%) and mouse IgM (12%). It has minimum cross-reactivity with immunoglobulins from other species. Reconstitute with 0.5 mL of distilled water (1.0 mg/mL after restoration).
Specifications
Specifications
| Antigen | Human IgM |
| Applications | ELISA, Immunocytochemistry, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide; pH 7.8 |
| Host Species | Goat |
| Purification Method | Affinity chromatography |
| Quantity | 500 μg |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.