Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Goat anti-Human Kappa Light Chain Secondary Antibody, Rhodamine, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP173598
Description
Kappa Light Chain Polyclonal specifically detects Kappa Light Chain in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.Specifications
Kappa Light Chain | |
Polyclonal | |
Rhodamine | |
HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 | |
Goat | |
Affinity Purified | |
RUO | |
3514 | |
Reconstitute with 1.0 ml deionized water (or equivalent). | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
LYOPH | |
Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
IGKC | |
Human kappa light chain | |
1 mg | |
Secondary | |
Specificity was confirmed by ELISA minimal cross reactivity against other human heavy or light chain isotypes. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction