Learn More
Bethyl Laboratories, Inc Goat anti-Human Kappa Light Chain Antibody HRP Conjugated, 1mg
Supplier:  Bethyl Laboratories, Inc A80115P
Encompass_Preferred
Reactivity Human, Applications: WB, IHC, ICC, ELISA; Host Goat, Antibody Type Polyclonal, Conjugate HRP, Format Whole IgG
Catalog No. NC9943754
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.