Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFT57, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89116792 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-792 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-792 Supplier Abnova Corporation Supplier No. PAB13672
Only null left
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of IFT57.

Sequence: C-EQPQEYDDPNATISN

Specifications

Antigen IFT57
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of IFT57.
Dilution ELISA (1:8000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene IFT57
Gene Alias ESRRBL1/FLJ10147/HIPPI/MHS4R2
Gene Symbols IFT57
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids at internal region of human IFT57.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55081
Test Specificity Approximately 49 KDa band observed in human and mouse brain lysates (calculated MW of 49.1 KDa according to human NP_060480.1 and of 48.8 KDa according to mouse NP_082956.2).
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.