Learn More
Alpha Diagnostic International Goat Anti-Monkey IgG (gamma-chain specific)-HRP conjugate | 0.5mL
SDP
Supplier:  Alpha Diagnostic International 70021
Encompass
Specificity has been tested using IEP, immunodiffusion, and ELISA. The product reacts with monkey IgG only. The antibody may recognize other species IgGs that have common gamma chains, particularly IgG's from other related monkeys. However, no antibody is detected to other serum proteins or IgM or IgA.
Catalog No. NC1863434
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.