Learn More
Description
Specifications
Specifications
| Antigen | IgG Heavy Chain |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | FITC |
| Formulation | PBS |
| Gene Alias | Immunoglobulin G, ImmunoglobulinG |
| Host Species | Goat |
| Immunogen | Goat anti-Monkey IgG (gamma chain) Secondary Antibody was produced by repeated immunization with monkey IgG gamma heavy chain in goat. |
| Purification Method | Multi-step |
| Quantity | 0.5 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
