Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Monkey IgG Heavy Chain Secondary Antibody, Novus Biologicals™
SDP

Catalog No. NBP260692 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP260692 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP260692 Supplier Novus Biologicals Supplier No. NBP260692
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG Heavy Chain Polyclonal specifically detects IgG Heavy Chain in Monkey samples. It is validated for ELISA, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG Heavy Chain
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, ELISA 1:75000 - 1:175000, Immunohistochemistry 1:1000-1:5000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Alias Immunoglobulin G, ImmunoglobulinG
Host Species Goat
Immunogen Goat anti-Monkey IgG (gamma chain) Secondary Antibody was produced by repeated immunization with monkey IgG gamma heavy chain in goat.
Purification Method Affinity Purified
Quantity 1 mg
Primary or Secondary Secondary
Test Specificity No reaction was observed against Monkey IgM or Monkey IgA.
Target Species Monkey
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.