Learn More
Carbosynth LLC. Goat anti Monkey IgM secondary antibody (HRP)
SDP
Supplier:  Carbosynth LLC. 43RIG074HRP
Encompass_Preferred
Immunoglobulin M, or IgM for short, is a basic antibody that is produced by B cells. It is the primary antibody against A and B antigens on red blood cells. IgM is by far the physically largest antibody in the human circulatory system. It is the first antibody to appear in response to initial exposure to antigen.
Catalog No. 50-125-3323
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.