Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Mouse Complement C3 Polyclonal Anitbody, MP Biomedicals

Catalog No. ICN55463 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN55463 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN55463 Supplier MP Biomedicals, Inc Supplier No. 0855463
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Host Species: Goat
Species Reactivity: Mouse

TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement C3
Classification Polyclonal
Conjugate Unconjugated
Host Species Goat
Quantity 5 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.