Learn More
Goat anti-Mouse IgA/IgG/IgM (H+L) Secondary Antibody, DyLight 350, R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgA/IgG/IgM (H+L) |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 350 |
| Formulation | 50 mM Sodium Borate |
| Host Species | Goat |
| Immunogen | Mouse IgG IgA and IgM whole molecule |
| Purification Method | Multi-step |
| Quantity | 0.5 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.