Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Mouse IgA/IgG/IgM (H+L) Secondary Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP172780C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP172780C 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP172780C Supplier Novus Biologicals Supplier No. NBP172780C
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgA/IgG/IgM (H+L) Polyclonal antibody specifically detects IgA/IgG/IgM (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen IgA/IgG/IgM (H+L)
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Host Species Goat
Immunogen Mouse IgG IgA and IgM whole molecule
Purification Method Multi-step
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.