Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Mouse IgG F(ab')2 Secondary Antibody, PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB000151 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB000151 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB000151 Supplier Novus Biologicals Supplier No. NB7588PECY550.5ML
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG F(ab)2 Polyclonal specifically detects IgG F(ab)2 in Mouse samples. It is validated for Dot Blot, ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot.

Specifications

Antigen IgG F(ab)2
Applications Dot Blot, ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate PE-Cyanine5.5
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot
Formulation PBS
Host Species Goat
Immunogen Mouse IgG F(ab')2 fragment
Purification Method Affinity Purified
Quantity 0.5 mL
Test Specificity Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Goat Serum, Mouse IgG, Mouse IgG F(ab)2 and Mouse Serum. No reaction was observed against Mouse IgG F(c).
Target Species Mouse
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.