Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Mouse IgG Fc Secondary Antibody (Pre-adsorbed), Novus Biologicals™
SDP

Catalog No. NBP261925 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP261925 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP261925 Supplier Novus Biologicals Supplier No. NBP261925
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG Fc Polyclonal specifically detects IgG Fc in Mouse samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG Fc
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:20000, ELISA 1:1000 - 1:20000, Immunohistochemistry 1:200 - 1:2000, Immunocytochemistry 1:200 - 1:2000
Formulation Phosphate Buffered Saline (PBS) with 0.09% Sodium Azide
Host Species Goat
Immunogen Mouse IgG
Purification Method Affinity Purified
Quantity 0.5 mg
Primary or Secondary Secondary
Test Specificity By immunoelectrophoresis and ELISA this antibody reacts specifically with mouse IgG. Cross reactivity with IgA and IgM is negligible. No antibody was detected against non-immunoglobulin serum proteins. Antiserum was cross adsorbed using human and rat immunosorbents to remove cross reactive antibodies. Less than 1% cross reactivity to human and rat IgG was detected.This antibody may cross react with IgG from other species.
Target Species Mouse
Content And Storage Store at 4C.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.