Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Mouse IgG (H+L) Secondary Antibody, DyLight 550 (Pre-adsorbed), Novus Biologicals™
SDP

Catalog No. NBP175956 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP175956 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP175956 Supplier Novus Biologicals Supplier No. NBP175956
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Flow Cytometry, Immunomicroscopy.

Specifications

Antigen IgG (H+L)
Applications Flow Cytometry, Immunomicroscopy
Classification Polyclonal
Concentration LYOPH
Conjugate DyLight 550
Dilution Flow Cytometry 1:20 - 1:2000, Immunomicroscopy 1:20 - 1:2000
Host Species Goat
Immunogen Purified Mouse IgG (H&L)
Purification Method Affinity purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity Based on IEP, this antibody reacts with heavy gamma chains on mouse IgG and light chains on all mouse immunoglobulins. This antibody has been pre-adsorbed against human IgG and serum proteins.
Reconstitution Rehydrate with 1.1 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily.
Target Species Mouse
Content And Storage Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.