Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Goat anti-Mouse IgG (H+L) Secondary Antibody, Janelia Fluor 549, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP268559JF549
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Mouse samples. It is validated for Flow Cytometry, ELISASpecifications
IgG (H+L) | |
Polyclonal | |
50mM Sodium Borate | |
Mouse IgG, whole molecule | |
0.5 mL | |
Adaptive Immunity, Immunology | |
Mouse | |
Purified |
Flow Cytometry, ELISA | |
Janelia Fluor 549 | |
Goat | |
Affinity purified | |
RUO | |
Secondary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction