Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Mouse IgG (H+L) Secondary Antibody, PE (Pre-adsorbed), R&D Systems™
SDP

Catalog No. NB7594 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB7594 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB7594 Supplier R&D Systems Supplier No. NB7594
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Goat Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Dot Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen IgG (H+L)
Applications Dot Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate PE
Dilution Flow Cytometry 1:100-1:250, Immunocytochemistry/Immunofluorescence 1:100-1:250, Dot Blot
Host Species Goat
Immunogen Goat anti-Mouse IgG (H+L) Secondary Antibody [PE] (Pre-adsorbed) was produced by repeated immunization with Mouse IgG whole molecule in goat.
Purification Method Affinity Purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity No reaction was observed against Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Rabbit, Rat and Sheep Serum Proteins.
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Product Type Antibody
Isotype IgG
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.