Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Mouse IgG (H+L) Secondary Antibody, PerCP, R&D Systems™
SDP

Catalog No. NB537840 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB537840 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB537840 Supplier R&D Systems Supplier No. NB7567PCP
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Goat Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen IgG (H+L)
Applications ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Host Species Goat
Immunogen This Goat anti-Mouse IgG (H+L) Secondary Antibody (Pre-adsorbed) was developed against mouse IgG .
Purification Method Immunogen affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.