Learn More
Description
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | ChIP Assay, Dot Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 8.6 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:3000 - 1:12000, ELISA 1:100000 - 1:500000, Immunohistochemistry 1:1000 - 1:6000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:5000 - 1:25000, Immunohistochemistry-Frozen 1:5000 - 1:25000, Dot Blot, Chromatin Immunoprecipitation (ChIP) |
| Host Species | Goat |
| Immunogen | Mouse IgG whole molecule |
| Purification Method | Affinity Purified |
| Quantity | 2 mg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
