Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Mouse IgG1 Heavy Chain Secondary Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NBP169914B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP169914B 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP169914B Supplier Novus Biologicals Supplier No. NBP169914B
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG1 Heavy Chain Polyclonal specifically detects IgG1 Heavy Chain in Mouse samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG1 Heavy Chain
Applications ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Biotin
Dilution Western Blot 1:10000- 1:200000, ELISA 1:10000 - 1:200000, Immunohistochemistry 1:250 - 1:2500, Immunocytochemistry/Immunofluorescence 1:100- 1:500
Host Species Goat
Immunogen Mouse IgG1
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity By immunoelectrophoresis and ELISA this antibody reacts specifically with mouse IgG1. Cross reactivity to mouse IgA, IgM, IgG2a, IgG2b, IgG2c, IgG3 and IgE is undetectable. Some hybridoma clones may express aberrant immunoglobulin-related peptides that are improperly recognized by this antibody.
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.